D-(-)-Sorbitol 91.0-100.5% (by anhydrous basis) NF, Multi-Compendial
Product Details & Documents
Sorbitol, Multi-Compendial.
Only class 2 solvents (ethylene glycol) are likely to be present. All are below option 1 limits.
- CAS Number:50-70-4
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00004708
- Molecular Weight:182.17 g/mol
- :Management of Change (MOC) category = R
Documents
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals
Product Literature
sds_BAKR3449
SDS
spec_JT3449-06 spec_JT3449-9
Product specification
Specifications
Specification Test Results
- NF - Assay (anhydrous basis):91.0 - 100.5 %
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - pH of 10% solution:3.5 - 7.0
- NF - Reducing Sugars:≤ 0.3 %
- NF - Residue on Ignition:≤ 0.1 %
- NF - Nickel (Ni):≤ 1 ppm
- NF - Chloride (Cl):≤ 0.0050 %
- NF - Sulfate (SO₄):≤ 0.01 %
- NF - Water:≤ 1.5 %
- NF - Total Aerobic Microbial Count, cfu/g:≤ 1000
- NF - Total Yeast and Mold Count, cfu/g:≤ 100
- EP/BP - Assay (anhydrous basis):97.0 - 102.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Conductivity, uS cm-1:≤ 20
- EP/BP - Reducing Sugars:≤ 0.2 %
- EP/BP - Related Substances - Any impurity, each:≤ 2.0 %
- EP/BP - Related Substances - Total:≤ 3.0 %
- EP/BP - Water:≤ 1.5 %
- Total Viable Aerobic Count - Total Aerobic Microbial Count, cfu/g:≤ 100
- Total Viable Aerobic Count - Total Yeast and Mold Count, cfu/g:≤ 100
- Total Viable Aerobic Count - Escherichia coli (negative):Passes Test
- Salmonella (negative):Passes Test
- Endotoxin Concentration, <2.5 IU/g:Passes Test
- JP - Assay (dried basis):≥ 97.0 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Identification C:Passes Test
- JP - Clarity and Color:Passes Test
- JP - Acidity or Alkalinity:Passes Test
- JP - Chloride (Cl):≤ 0.005 %
- JP - Sulfate (SO₄):≤ 0.006 %
- JP - Heavy Metals (as Pb):≤ 5 ppm
- JP - Nickel (Ni):Passes Test
- JP - Arsenic (As):≤ 1.3 ppm
- JP - Glucose:Passes Test
- JP - Sugars:Passes Test
- JP - Loss on Drying:≤ 2.0 %
- JP - Residue on Ignition:≤ 0.02 %
Specifications
- Pack type:Drum, blue, open head
- Size:25 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



