Order Entry
Canada
Orders LinkContactUsLinkComponent
Macron Fine Chemicals™ BuffAR®, pH Buffer Solutions

Macron Fine Chemicals™ BuffAR®, pH Buffer Solutions

±0.01, PE bottle, 500 ml (16.9 oz.), 4.01, Red
Catalog # CAMK009704
Supplier #:  MACR0097-04
Macron Fine Chemicals™ BuffAR®, pH Buffer Solutions
Catalog # CAMK009704
CAS Number:  
MOC:  Non-Regulated, With InformationCertificates
Macron Fine Chemicals™ BuffAR®, pH Buffer Solutions
Macron Fine Chemicals™ BuffAR®, pH Buffer Solutions

±0.01, PE bottle, 500 ml (16.9 oz.), 4.01, Red

CAS Number:  
MOC:  Non-Regulated, With Information
Price
$168.83
Log in for availability
Each(1/Itm/Bx) Case of 12 (1/ Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Specifications

Specifications

  • Pack type:
    PE bottle
  • Color:
    Red
  • Size:
    500 ml (16.9 oz.)
  • pH:
    4.01
  • pH accuracy:
    ±0.01

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.