Eisco® Individual Bones
Tibia Bone, Left Side
470308-078
: Wards
: AMCH1029AS
For Individual Study.
- Sold in left and right side variants
- Great addition for your current skeleton
These individual bones are a great addition to you collection. They can be used as a match for your existing Eisco disarticulated skeleton, or to allow students to compare general skeletal structure. Additionally, these can be used as additional bones to simulate forensics activities, and skeletal reconstruction labs. Unless otherwise indicated, bones are not prepared in an articulated form, and have not been drilled.
- Description:Tibia Bone, Left Side