Orders LinkContactUsLinkComponent
pBlueScript II Vectors, Agilent Technologies
pBlueScript II Vectors, Agilent Technologies
pBluescript II SK(–) Phagemid Kit, 1 Kit, f1 origin in (−) orientation, Sac-Kpn, polylinker orientation. Contains: 20 μg pBluescript II SK(−) phagemid vector, host strain: XL1-Blue MRF'
Catalog # CA99900-342
pBlueScript II Vectors, Agilent Technologies
Catalog # CA99900-342
Supplier:  
CAS Number:  
pBlueScript II Vectors, Agilent Technologies
pBlueScript II Vectors, Agilent Technologies
pBluescript II SK(–) Phagemid Kit, 1 Kit, f1 origin in (−) orientation, Sac-Kpn, polylinker orientation. Contains: 20 μg pBluescript II SK(−) phagemid vector, host strain: XL1-Blue MRF'
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The pBluescript II phagemids (plasmids with a phage origin) are cloning vectors designed to simplify commonly used cloning and sequencing procedures, including the construction of nested deletions for DNA sequencing, generation of RNA transcripts in vitro and site-specific mutagenesis and gene mapping.
  • High copy number ColE1-based phagemid
  • Large and versatile polylinker in two orientations, f1 origin available in either (+) or (–) orientation
  • T3 and T7 promoters for in vitro transcription of RNA
The pBluescript II phagemids have an extensive polylinker with 21 unique restriction enzyme recognition sites.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Description:
    pBluescript II SK(–) Phagemid Kit
  • Size:
    1 Kit
  • Details:
    f1 origin in (−) orientation, Sac-Kpn, polylinker orientation. Contains: 20 μg pBluescript II SK(−) phagemid vector, host strain: XL1-Blue MRF'
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)