Order Entry
Canada
Orders LinkContactUsLinkComponent
Universal Reverse Primer, Agilent Technologies

Universal Reverse Primer, Agilent Technologies

Universal reverse primer, 50
Catalog # CA97066-434
Supplier #:  600037
Universal Reverse Primer, Agilent Technologies
Catalog # CA97066-434
CAS Number:  
Universal Reverse Primer, Agilent Technologies
Universal Reverse Primer, Agilent Technologies

Universal reverse primer, 50

CAS Number:  
Price
$218.00
Log in for availability
Each (200µl)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The universal reverse primer serves as the downstream primer in the qPCR reaction by annealing to a universal sequence tag that is added to the cDNA during the reverse transcription protocol.

Applications: PCR
Number of Reactions: 3.125 μM
Physical Form: Liquid
Reactions per Kit: 300
Volume: 200 µL

Documents

Specifications

Specifications

  • Description:
    Universal reverse primer
  • No. of reactions:
    50

Specifications