pBlueScript II Vectors, Agilent Technologies
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The pBluescript II phagemids (plasmids with a phage origin) are cloning vectors designed to simplify commonly used cloning and sequencing procedures, including the construction of nested deletions for DNA sequencing, generation of RNA transcripts in vitro and site-specific mutagenesis and gene mapping.
- High copy number ColE1-based phagemid
- Large and versatile polylinker in two orientations, f1 origin available in either (+) or (–) orientation
- T3 and T7 promoters for in vitro transcription of RNA
The pBluescript II phagemids have an extensive polylinker with 21 unique restriction enzyme recognition sites.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
- Description:pBluescript II XR Predigested Vector
- Size:55 Reactions
- Details:XR vector is predigested with Xho I and EcoR I
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.