Multiple Affinity Removal Starter Reagent Kit, Agilent Technologies
Spin filters for Gel Matrix, For sample pre-cleanup, cellulose acetate, 0.22 µm
Catalog # CA76204-920
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
: 5185-5990
Multiple Affinity Removal Starter Reagent Kit, Agilent Technologies
Catalog # CA76204-920
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Starter Reagent Kit for Multiple Affinity Removal LC Columns.
: Includes: 2 bottles Buffer A (1 l), 1 bottle Buffer B (1 l), 2 packs of 25 spin filters, 1 pack of 25 spin concentrator
Specifications
Specifications
- Description:Spin filters for Gel Matrix
- Size:0.22 µm
- Details:For sample pre-cleanup, cellulose acetate



