Laboratory Warning Labels
Product Details & Documents
Various warning and safety labels for use throughout the lab. Designed to inform personnel of the multiple hazards present in the lab, including the presence of radioactive material, cytotoxic compounds, and biological waste.
- Temperature range: ambient temperature
- Removable or permanent adhesive
- All labels are preprinted
Multi-purpose warning labels, ideal for offices, clinics, and laboratories. Paper warning labels feature a permanent adhesive to ensure they won't wrinkle or peel, and are designed for use at ambient temperatures. These general-use safety labels are fully customizable with logos, images, and alphanumeric text, in order to display appropriate warning information.
Specifications
- Color:Red/white on black
- Description:Warning labels - CYTOTOXIC
- Dimensions:25.4×38 mm (1×1.5")
- Packaging:1000 labels/roll (1 label across)
- Core size:3"
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



