Orders LinkContactUsLinkComponent
Injection Training Pads
Injection Training Pads
Epi-Layers, —, —, —
Catalog # 470306-772
Supplier #:  REM-INJ-EPI-01
Injection Training Pads
Catalog # 470306-772
Supplier:  
Injection Training Pads
Injection Training Pads
Epi-Layers, —, —, —
Discontinued
Product Details & Documents
Remedy's medical training injection pads are designed for the practice of needle injections.
The pads can be used to simulate intradermal, intra-muscular and subcutaneous injections. The larger pad allows you to have more practice area, as well as practicing Z-track injections. A removable epidermis layer is placed on the surface of the pad allowing for the simulation of a "wheal" during intradermal injections.
The skin puncture resistance, as well as the resistance throughout the injection, closely mimics the feel of a real injection. Both pads have drainage on the bottom so they can be used with fluid filled syringes.
Ordering information: Additional epidermis layers may be purchased separately.
Specifications
  • Description:
    Epi-Layers
  • Height:
  • Length:
  • Width:
Recommendations