Injection Training Pads
Product Details & Documents
Remedy's medical training injection pads are designed for the practice of needle injections.
The pads can be used to simulate intradermal, intra-muscular and subcutaneous injections. The larger pad allows you to have more practice area, as well as practicing Z-track injections. A removable epidermis layer is placed on the surface of the pad allowing for the simulation of a "wheal" during intradermal injections.
The skin puncture resistance, as well as the resistance throughout the injection, closely mimics the feel of a real injection. Both pads have drainage on the bottom so they can be used with fluid filled syringes.
Ordering information: Additional epidermis layers may be purchased separately.
Specifications
- Description:Large Injection Pad "Patrick"
- Height:1.75"
- Length:6"
- Width:6"
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



