Order Entry
Canada
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid ACS

L(+)-Ascorbic acid ACS

Plastic bottle, 500 g
Catalog # RCRDCA075500B1
Supplier:   Ricca Chemical
Supplier #:  RDCA0750-500B1
L(+)-Ascorbic acid ACS
Catalog # RCRDCA075500B1
Supplier:  Ricca Chemical
CAS Number:  50-81-7
L(+)-Ascorbic acid ACS
L(+)-Ascorbic acid ACS

Plastic bottle, 500 g

CAS Number:  50-81-7
Price
1 142,82 $
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Synonyms:  Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Assay:
    min 99.0 %
  • Specific rotation:
    20.5-21.5 °
  • Heavy Metals (as Pb):
    max 0.002 %
  • Iron (Fe):
    max 0.001 %
  • Residue after Ignition:
    max 0.1 %

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.