FectoCHO® Expression System
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
FectoCHO® Expression system consists in the synergistic association of a novel CHO chemically defined medium, FectoCHO® CD Expression medium and a powerful transfection reagent, FectoPRO®.
- Outstanding protein production yields in various CHO cells
- Fast and effortless CHO cell adaptation
- Chemically defined kit components
- Excellent lot to lot reproducibility
- Cost-effective solution
FectoCHO® CD Expression medium allows easy cultivation of various strains of CHO cells such as CHO-K1, CHO-S and ExpiCHO®-S , without the need of an extensive sequential adaptation. FectoPRO®-mediated transient transfection in this chemically defined medium leads to the highest protein and antibody yields ever reached using Transient Gene Expression, up to 4 times higher than with the competitor's systems.
Certifications: Since 2002, Polyplus-transfection® is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Ordering information: FectoPRO® transfection reagent, FectoPRO® Booster and FectoCHO® CD Expression Medium can be purchased separately.
Documents
Polyplus-transfection® Protein and mAb Production Enhanced Transient Protein Production in CHO.pdf
Product Literature
Poster FectoCHO Expression System.pdf
Application Notes
Specifications
Specifications
- Description:FectoCHO™ Expression System
- Size:1 mL (Transfection Reagent), 1 L (Expression Medium)
- Buffer volume:1 ml
- Cat. No.:CA76234-736
- Cell specific:CHO
- Bioproduction:Yes
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



