Orders LinkContactUsLinkComponent
Synthware Round-Bottom Flasks with Three Necks, Angled
3000 ml, 24/40, 24/40
Catalog # 76264-080
Supplier:   Kemtech America
 :  F43243L
Synthware Round-Bottom Flasks with Three Necks, Angled
Catalog # 76264-080
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
3000 ml, 24/40, 24/40
Price
413,39 $
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Round bottom with [ST] joints.
  • 3 Angled necks
  • Ground glass joints
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Specifications
  • Capacity:
    3000 ml
  • Center neck [ST] joint:
    24/40
  • Side neck ST joints:
    24/40
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.