Order Entry
Canada
Orders LinkContactUsLinkComponent
Storage Bins, Shaefer Systems

Storage Bins, Shaefer Systems

EF Stackable Conductive Container Solid Base/Sides, Black, 12W×16L×13H"
Catalog # 76248-700
Supplier #:  EF4320.PP3
Storage Bins, Shaefer Systems
Catalog # 76248-700
Storage Bins, Shaefer Systems

EF Stackable Conductive Container Solid Base/Sides, Black, 12W×16L×13H"

Price
$341.36
Log in for availability
Each
Product Details & Documents

Schaefer Shelving complete line of Conductive Containers protect sensitive electronic components from electrostatic fields or induced fields during production, shipping and storage.

  • All containers are returnable, reusable and recyclable
  • Molded with conductive carbon-filled polypropylene
  • Double-wall construction for long term performance
  • Recessed grip handle on back for easier manual handling

Permanently conductive and unaffected by washing with normal cleaners. These containers are molded with a conductive carbon-filled polypropylene material to provide a barrier which these fields cannot penetrate.

Specifications
  • Color:
    Black
  • Description:
    EF Stackable Conductive Container Solid Base/Sides
  • Dimensions:
    12W×16L×13H"