Order Entry
Canada
Orders LinkContactUsLinkComponent
Storage Bins, Shaefer Systems

Storage Bins, Shaefer Systems

Stackable Conductive Lid, Black, 16W×24L"
Catalog # 76244-980
Supplier #:  EFD64.PP3
Storage Bins, Shaefer Systems
Catalog # 76244-980
Storage Bins, Shaefer Systems

Stackable Conductive Lid, Black, 16W×24L"

Price
$165.53
Log in for availability
Each
Product Details & Documents

Schaefer Shelving complete line of Conductive Containers protect sensitive electronic components from electrostatic fields or induced fields during production, shipping and storage.

  • All containers are returnable, reusable and recyclable
  • Molded with conductive carbon-filled polypropylene
  • Double-wall construction for long term performance
  • Recessed grip handle on back for easier manual handling

Permanently conductive and unaffected by washing with normal cleaners. These containers are molded with a conductive carbon-filled polypropylene material to provide a barrier which these fields cannot penetrate.

Specifications
  • Color:
    Black
  • Description:
    Stackable Conductive Lid
  • Dimensions:
    16W×24L"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.