VWR® Cap Mat
96-Well Round Well Cap Mat, EVA, Sterile
Supplier: VWR International
Jump to:
Product Details & Documents
Re-usable cap mats are available for friction sealing of plates.
Cap mats are an economic alternative to thermal or adhesive seals as they can be sterilized and reused.
Documents
Specifications
- Sterility:Sterile
- Description:96-Well Round Well Cap Mat
- Material:EVA