Swingsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Product Details & Documents
Secure specimens during processing with the Swingsette™ Tissue Processing/Embedding Cassette, in Quickload™ stacks.
- In Quickload™ stacks
- 45° anterior writing area
- Easy to assemble hinged lids
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Specifications
- Color:Gray
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)