Orders LinkContactUsLinkComponent
Slimsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport | Avantor
Slimsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Aqua
Catalog # CA76214-142
Supplier:   Simport Scientific
Fournisseur #:  M509-12T
Slimsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Catalog # CA76214-142
Supplier:  Simport Scientific
Slimsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Slimsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Aqua
Price
$1,404.18
Log in for availability
Case of 2,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Secure specimens during processing with the Slimsette™ Tissue Processing/Embedding Cassette, in Quickload™ stacks.
  • In Quickload™ stacks
  • 45° anterior writing area
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes.
Specifications
  • Color:
    Aqua
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.