Orders LinkContactUsLinkComponent
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport | Avantor
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Orange
Catalog # CA76214-106
Supplier:   Simport Scientific
Fournisseur #:  M505-11T
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Catalog # CA76214-106
Supplier:  Simport Scientific
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Orange
Price
$394.35
Log in pour la disponibilité
Case of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Secure specimens during processing with the Unisette™ Tissue Processing/Embedding Cassette.
  • In Quickload™ stacks
  • 35° Anterior writing area
  • One-piece construction with hinge-lock design prevents early separation of base and lid
  • Snap latch and hinge-lock design; cassettes will always relock securely
  • Allows for one-hand operation
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
The snap latch and hinge-lock design allows for one-hand operation and always relocks securely without danger of specimen loss.
Certifications: CE-IVD.
Packaging: Each case contains 25 Quickload™ stacks of 40 cassettes.
Spécifications
  • Color:
    Orange
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.