Histosette® II Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Product Details & Documents
Secure specimens during processing with the Histosette® II Tissue Processing/Embedding Cassette.
- In Quickload™ stacks
- 45° anterior writing area
- Open and close as often as necessary; cassettes will always relock securely
- Back-mounted tab allowing an unobstructed anterior marking area
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Keep specimens safely submerged in liquid. The flow-through slots maximize fluid exchange. The one-piece integral lid eliminates the need for separate steel lids.
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes.
Specifications
- Color:White
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.