SureVector, Vectors, Agilent Technologies
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Combine standard DNA components to build custom vector constructs from Agilent’s set of standard parts.
- Rapid custom vector generation - Less than a day from design to vector, compared to 4 weeks for custom vector services
- Reliable and precise assembly - SureVector is extensively validated to ensure that standard parts can be interchanged, without loss of functionality
- More flexible than traditional systems - Assemble new vectors in your lab as experimental requirements change, versus always ordering a new one
- Control your experiments - Take control of your experiments, troubleshoot your own work, not that of your service providers DNA assembly
SureVector next-generation cloning uses the power of synthetic biology to combine standard DNA components to build custom vector constructs from the Agilent set of standard parts. Simply design a vector, order or PCR your gene-of-interest, and your custom plasmid is just a single reaction away.
Proprietary enzymes enable rapid and reliable assembly of multiple DNA fragments into a circular, recombinant plasmid containing your target gene.
Delivery information: Components include: bacterial selection markers, bacterial origins of replication, mammalian selection markers, promoters for yeast, mammalian cells and bacteria, and a variety of solubility/affinity tags; Enabling functionality in a range of cells.
Caution: For research use only. Not for use in diagnostic procedures.
Documents
Specifications
Specifications
- Description:SureVector Mammalian C-terminal expansion kit
- Size:15 reactions
- Details:Promoters and C-terminal tags for expression in mammalian cells
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)