Orders LinkContactUsLinkComponent
SureVector, Vectors, Agilent Technologies
SureVector, Vectors, Agilent Technologies
SureVector E. coli N-terminal expansion kit, 15 reactions, SureVector E. coli N-terminal Expansion Kit expands functionality of the core kit in E. coli by including additional promoters and N-terminal tags
Catalog # CA76193-664
SureVector, Vectors, Agilent Technologies
Catalog # CA76193-664
Supplier:  
CAS Number:  
SureVector, Vectors, Agilent Technologies
SureVector, Vectors, Agilent Technologies
SureVector E. coli N-terminal expansion kit, 15 reactions, SureVector E. coli N-terminal Expansion Kit expands functionality of the core kit in E. coli by including additional promoters and N-terminal tags
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Combine standard DNA components to build custom vector constructs from Agilent’s set of standard parts.
  • Rapid custom vector generation - Less than a day from design to vector, compared to 4 weeks for custom vector services
  • Reliable and precise assembly - SureVector is extensively validated to ensure that standard parts can be interchanged, without loss of functionality
  • More flexible than traditional systems - Assemble new vectors in your lab as experimental requirements change, versus always ordering a new one
  • Control your experiments - Take control of your experiments, troubleshoot your own work, not that of your service providers DNA assembly
SureVector next-generation cloning uses the power of synthetic biology to combine standard DNA components to build custom vector constructs from the Agilent set of standard parts. Simply design a vector, order or PCR your gene-of-interest, and your custom plasmid is just a single reaction away.
Proprietary enzymes enable rapid and reliable assembly of multiple DNA fragments into a circular, recombinant plasmid containing your target gene.
Delivery information: Components include: bacterial selection markers, bacterial origins of replication, mammalian selection markers, promoters for yeast, mammalian cells and bacteria, and a variety of solubility/affinity tags; Enabling functionality in a range of cells.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Description:
    SureVector E. coli N-terminal expansion kit
  • Size:
    15 reactions
  • Details:
    SureVector E. coli N-terminal Expansion Kit expands functionality of the core kit in E. coli by including additional promoters and N-terminal tags
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)