Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Gradient Buffers

pH Gradient Buffers

CX-1 pH gradient buffer A, 500 ml, 5.6
Catalog # 76205-170
Supplier:   Thermo Scientific
Supplier #:  302779
pH Gradient Buffers
Catalog # 76205-170
Supplier:  Thermo Scientific
CAS Number:  
pH Gradient Buffers

CX-1 pH gradient buffer A, 500 ml, 5.6

CAS Number:  
Price
$1,188.00
New Customer Promotional Price
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Deliver robust, reproducible gradients with the Thermo Scientific pH buffer platform, which is applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.

  • Proprietary buffer formulations enable fast, robust and reproducible pH gradients that are simple to optimise and easily automated
  • Ready to use with existing LC columns and systems, without the need for time-consuming mobile phase adjustments
  • Applicable to the majority of MAbs

The pH gradient platform saves time in method development and facilitates method transfer to QA/QC for a wide range of MAb charge variants through a generic LC-based approach to MAb characterization.

All that is required to generate a pH gradient is a 1:10 dilution of the pH buffer concentrates. A linear pH gradient from pH 5.6 to 10.2 can simply be delivered by running a pump gradient from 100% eluent A to 100% eluent B.

Specifications

Specifications

  • Description:
    CX-1 pH gradient buffer A
  • Size:
    500 ml
  • pH:
    5.6

Specifications