CRITERION™ Phosphate Buffer, pH 7.2
10 kg, Bucket
: C8333
CRITERION™ Agar, bacteriological grade is used as a solidifying agent in the preparation of solid and semi-solid culture media.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- Increased solubility - dissolves quickly
- Custom and bulk products available - to your specifications
- BSE-free
- Pack type:Bucket
- Size:10 kg
- Application:Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing
- Cat. No.:CA76196-446
- Storage temperature:2...30 °C



