Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Certified Reference Materials, Oxide

VWR® Certified Reference Materials, Oxide

2 kg, Gold, 0 - 10 / > 1000 ppm, Au/Ag, Oxide, Jar
Catalog # CA76202-570
Supplier #:  21002141
VWR® Certified Reference Materials, Oxide
Catalog # CA76202-570
CAS Number:  
VWR® Certified Reference Materials, Oxide
VWR® Certified Reference Materials, Oxide

2 kg, Gold, 0 - 10 / > 1000 ppm, Au/Ag, Oxide, Jar

CAS Number:  
Price
$140.59
Log in for availability
Each (2,000g) Case of 4 (2,000,g)
Product Details & Documents

KLEN's CRMs are certified products containing a range of analytes including Au, Ag, Pt, and Pd at various ppm levels

Ordering information: Other sizes (30g, 50g and 150g sachets) are available, please contact your Avantor sales representative for more information.

Specifications

Specifications

  • Concentration:
    0 - 10 / > 1000 ppm
  • Matrix:
    Oxide
  • Pack type:
    Jar
  • Size:
    2 kg
  • Analyte:
    Au/Ag
  • Element:
    Gold

Specifications