Order Entry
Canada
Orders LinkContactUsLinkComponent
Wire Triangles

Wire Triangles

Side length: 51 mm, 26 mm
Catalog # 470235-760
Supplier:   GSC International
Supplier #:  4-2020-CS
Wire Triangles
Catalog # 470235-760
Supplier:  GSC International
Wire Triangles
Wire Triangles

Side length: 51 mm, 26 mm

Price
$43.75
Log in for availability
Pack of 10 Case of 100
Product Details & Documents

Perfect tool for suspending a crucible.

  • Uniform clay tubes
  • Galvanized iron wire resists rusting
Specifications
  • Ø:
    26 mm
  • Description:
    Side length: 51 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.