Orders LinkContactUsLinkComponent
Quality Corks, XX
Quality Corks, XX
XX Quality Corks, Size 3
Catalog # 470235-744
Supplier:   GSC International
Supplier #:  CS-3-100-10
Quality Corks, XX
Catalog # 470235-744
Supplier:  GSC International
Quality Corks, XX
Quality Corks, XX
XX Quality Corks, Size 3
Price
$22.30
Log in for availability
Pack of 100 Case of 1000
Product Details & Documents
Size Top Diameter Bottom Diameter
Size 1 11.1 mm 8.3 mm
Size 2 12.7 mm 9.5 mm
Size 3 14.2 mm 10.7 mm
Size 4 15.8 mm 11.9 mm
Size 6 19.0 mm 14.6 mm
Size 20 41.2 mm 34.0 mm
  • Size 2 - sold in a case; 10 packs of 100
  • Size 3 - sold in a case; 10 packs of 100
  • Size 4 - sold in a case; 10 packs of 100
  • Size 20 - sold in a pack of 10
  • The natural cork is safe for use with food products
Get the perfect fit with our lab-grade cork stoppers. These stoppers ensure a secure, leak-proof seal every time. Crafted from natural cork, these stoppers are safe for use with food products, making them a reliable choice for all your storage needs.
Specifications
  • Description:
    XX Quality Corks, Size 3
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.