Pulleys
Pulley, Triple Sheave
Catalog # 470235-736
Supplier: GSC International
Supplier #: 4-16063-15
Jump to:
Product Details & Documents
These free-running plastic pulleys are durable in design.
- Plastic Construction
- Durable
- Nickel-Plated Steel Blocks with Hooks
The sheaves are 50 mm in Ø and made of rigid plastic.
The nickel-plated steel blocks have hooks at the top and bottom.
Specifications
- Description:Pulley, Triple Sheave