Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BIN STACK HANG PP GREEN 16 X11 X 8 IN
Catalogue # 76202-864
Fournisseur:
QUANTUM STORAGE SYSTEMS
Agrandir l'image
BIN STACK HANG PP GREEN 16 X11 X 8 IN
Catalogue # 76202-864
Fournisseur:
QUANTUM STORAGE SYSTEMS
Supplier Number:
QUS255GN
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Fournisseur: VWR International
VWR® Disposable Petri Dishes
Fournisseur: VWR International
VWR® Nitrile Examination Gloves, Blue
Fournisseur: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Fournisseur: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
VWR® Universal Pipette Tips
Fournisseur: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma