Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Certified Reference Materials, Oxide

VWR® Certified Reference Materials, Oxide

2 kg, Gold, 0 - 10 /<100 ppm, Au/Ag, Oxide, Jar
Catalog # CA76202-550
Supplier #:  21002121
VWR® Certified Reference Materials, Oxide
Catalog # CA76202-550
CAS Number:  
VWR® Certified Reference Materials, Oxide
VWR® Certified Reference Materials, Oxide

2 kg, Gold, 0 - 10 /<100 ppm, Au/Ag, Oxide, Jar

CAS Number:  
Price
$150.38
Log in for availability
Each (2,000g) Case of 4 (2,000,g)
Product Details & Documents

KLEN's CRMs are certified products containing a range of analytes including Au, Ag, Pt, and Pd at various ppm levels

Ordering information: Other sizes (30g, 50g and 150g sachets) are available, please contact your Avantor sales representative for more information.

Specifications

Specifications

  • Concentration:
    0 - 10 /<100 ppm
  • Matrix:
    Oxide
  • Pack type:
    Jar
  • Size:
    2 kg
  • Analyte:
    Au/Ag
  • Element:
    Gold

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.