Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Mini Electrophoresis Systems

VWR® Mini Electrophoresis Systems

VWR® Mini Electrophoresis System, 115 V, 4.5×6.25×2.25"
Catalog # 76196-486
Supplier:   VWR International
VWR® Mini Electrophoresis Systems
Catalog # 76196-486
Supplier:  VWR International
VWR® Mini  Electrophoresis Systems
VWR® Mini Electrophoresis Systems

VWR® Mini Electrophoresis System, 115 V, 4.5×6.25×2.25"

Price
$1,235.03
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

VWR® Mini electrophoresis system is an ideal tool for quick and efficient nucleic acid separations.

  • Complete and convenient system
  • Power supply connects to the gel box, no wires are needed

This system is used for quick nucleic acid separations for a range of applications including evaluation of PCR results. The power supply has 3 voltage choices, connects to the gel tank so no wire leads are needed, and the running tank is compact and does not require a large amount of buffer.

Delivery information: Includes a power supply, gel tank, and gel casting stands, trays and combs, two gel casting sets with combs to make 10.5×6 cm and 5×6 cm gels.

Specifications
  • Description:
    VWR® Mini Electrophoresis System, 115 V
  • L×W×H:
    4.5×6.25×2.25"
  • Cat. No.:
    76196-486
  • Max. power:
    40 W
  • Voltage:
    35, 50 or 100 V (output)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.