Order Entry
Canada
Orders LinkContactUsLinkComponent
Packaging Buckets, Burkle

Packaging Buckets, Burkle

PP packaging bucket, 5 L (1.3 gal.), 225 mm (8.86"), 194 mm (7.6") with cap
Catalog # 76193-760
Supplier:   BURKLE INC
Supplier #:  2327-0050
Packaging Buckets, Burkle
Catalog # 76193-760
Supplier:  BURKLE INC
Packaging Buckets, Burkle
Packaging Buckets, Burkle

PP packaging bucket, 5 L (1.3 gal.), 225 mm (8.86"), 194 mm (7.6") with cap

Price
$14.40
Log in for availability
Each
Product Details & Documents

Packaging buckets for transport, packaging, and storing of various materials

  • Polypropylene, white finish

  • Air-tight closure
  • Supplied with lid
  • Autoclavable
  • Also available with advertising print (in large quantities on request)

Suitable for the food or building material industries and non-aggressive chemicals in environmental protection, hospitals, and the household. Buckets are equipped with a strong handle and a quality closure for product safety.

Specifications
  • Capacity:
    5 L (1.3 gal.)
  • Description:
    PP packaging bucket
  • Height:
    194 mm (7.6") with cap
  • Diameter:
    225 mm (8.86")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.