Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Prepared Agarose

Prepared Agarose Gel, 2.0%, 200 ml Bottle
Catalog # 470233-588
  
Supplier:   Ward's Science
 :  AA0082-200ML
Prepared Agarose
Catalog # 470233-588
Supplier:  Ward's Science
  
Prepared Agarose
Prepared Agarose

Prepared Agarose Gel, 2.0%, 200 ml Bottle

Price
40,90 $
Log in for availability
Each (200ml)
 
Product Details & Documents

Prepared with TBE buffer, these gels are easy to use — simply melt them in a microwave oven or water bath then pour them into a casting tray. The non-reusable gels have a one-year shelf life and should be stored at room temperature.

Specifications
  • Description:
    Prepared Agarose Gel, 2.0%, 200 ml Bottle
  • Cat. No.:
    470233-588