Order Entry
Canada
Orders LinkContactUsLinkComponent
Ampicillin | Avantor

Ampicillin

Ampicillin, Laboratory, 5 g
Catalog # 470233-552
Supplier:   Ward's Science
Fournisseur #:  AA0356-5G
Ampicillin
Catalog # 470233-552
Supplier:  Ward's Science
CAS Number:  
Ampicillin
Ampicillin

Ampicillin, Laboratory, 5 g

CAS Number:  
Price
$102.05
Log in for availability
Each (5g)
La quantité minimale de commande est de 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Add to prepared agar or broth to select antibiotic-resistant bacterial strains.

Shelf Life (months): 36
Storage: Green

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Ampicillin
  • Size:
    5 g
  • Chemical/physical specifications:
    Powder
  • Grade:
    Laboratory

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.