Orders LinkContactUsLinkComponent
Thread Adapters, Polypropylene
External thread: US Drum thread, Internal thread: 2" BSP, Silver
Catalog # 76077-152
Supplier:   BURKLE INC
 :  0600-1007
Thread Adapters, Polypropylene
Catalog # 76077-152
Supplier:  BURKLE INC
Thread Adapters, Polypropylene
Thread Adapters, Polypropylene
External thread: US Drum thread, Internal thread: 2" BSP, Silver
Price
76,34 $
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Bürkle offers a broad range of thread adapters to facilitate secure connections between many Bürkle pumps and discharge cocks to a variety of containers
This wide variety of adapters ensures users can find the correct connection.
Specifications
  • Color:
    Silver
  • Description:
    External thread: US Drum thread, Internal thread: 2" BSP