Thread Adapters, Polypropylene
External thread: 61 mm (2.40"), Internal thread: 2" BSP, Grey
Catalog # 76077-120
Supplier: BURKLE INC
Supplier #: 0600-1008
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Bürkle offers a broad range of thread adapters to facilitate secure connections between many Bürkle pumps and discharge cocks to a variety of containers
This wide variety of adapters ensures users can find the correct connection.
Specifications
- Color:Grey
- Description:External thread: 61 mm (2.40"), Internal thread: 2" BSP
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



