$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
VWR VERTICALRACK4X2 FOR 50C 1.5ML BOXES
76052-832
:
CRYSTAL TECHNOLOGY & INDUSTRIES
VWR VERTICALRACK4X2 FOR 50C 1.5ML BOXES
76052-832
:
CRYSTAL TECHNOLOGY & INDUSTRIES
:
CFM-415
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International