Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VWR UPRIGHT RACK1X4X2FOR 50C 1.5ML BOX
Catalog # 76052-726
Supplier:
CRYSTAL TECHNOLOGY & INDUSTRIES
Expand image
VWR UPRIGHT RACK1X4X2FOR 50C 1.5ML BOX
Catalog # 76052-726
Supplier:
CRYSTAL TECHNOLOGY & INDUSTRIES
Supplier Number:
UFM-1415
Frequently Bought Together
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International