Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VWR VERTICALRACK5X2 FOR 50C 0.5ML BOXES
Catalog # 76052-718
Supplier:
CRYSTAL TECHNOLOGY & INDUSTRIES
Expand image
VWR VERTICALRACK5X2 FOR 50C 0.5ML BOXES
Catalog # 76052-718
Supplier:
CRYSTAL TECHNOLOGY & INDUSTRIES
Supplier Number:
CFM-505
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International