Order Entry
Canada
Orders LinkContactUsLinkComponent
SpillStopper™ Work Surfaces for Protector® ClassMate™ Laboratory Hoods, Labconco®

SpillStopper™ Work Surfaces for Protector® ClassMate™ Laboratory Hoods, Labconco®

5 ft., Left rear, 150 lbs.
Catalog # 76045-670
Supplier:   Labconco
Supplier #:  9600510
SpillStopper™ Work Surfaces for Protector® ClassMate™ Laboratory Hoods, Labconco®
Catalog # 76045-670
Supplier:  Labconco
SpillStopper™ Work Surfaces for Protector® ClassMate™ Laboratory Hoods, Labconco®
SpillStopper™ Work Surfaces for Protector® ClassMate™ Laboratory Hoods, Labconco®

5 ft., Left rear, 150 lbs.

Price
$5,090.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Solid epoxy work surfaces are 3.2cm (11/4") thick, 65.7cm (257/8") deep, and dished to contain spills.

Solid epoxy cupsink cover (model 4005201; sold separately) covers cupsink when not in use.

Specifications
  • Shipping weight:
    150 lbs.
  • Width:
    5 ft.
  • CuPSInk cutout:
    Left rear
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Oval Epoxy Cupsink
    Oval Epoxy Cupsink
    30181-070
    Log in for availability
    Each - $202.00

Product Information

  • Catalog #
    Availability
    Price
  • Cupsink Cover
    Cupsink Cover
    97006-430
    Log in for availability
    Each - $116.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.