BOVINE FIBRINOGEN 90% CLOTTABLE WHT 5G
Catalog # CA76046-158
Supplier: MP Biomedicals
BOVINE FIBRINOGEN 90% CLOTTABLE WHT 5G
Catalog # CA76046-158
Supplier: MP Biomedicals
Supplier Number: 08820224
Frequently Bought Together
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Supplier: Thermo Fisher Scientific
Bulk Syringes, Nonsterile, BD Medical
Supplier: BD
Leakbuster™ Specimen Containers
Supplier: Starplex
Masterflex® Transfer Tubing, C-Flex®, Clear, Avantor®
Supplier: VWR International
VWR® Tissue Culture Dishes, Non-Treated, Sterilized, Non-Pyrogenic
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Masterflex® Union Fittings, Hose Barb, Tee, Polypropylene, Avantor®
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Agrobacterium tumafaciens
Supplier: Avantor
DNA MINICOLUMNS V-SPIN HIBIND PK1000
Supplier: Omega Bio-Tek
Hexacarbonylmolybdenum 98%
Supplier: Thermo Scientific Chemicals
Square Petri Dish, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Pierce Zeba™ Desalt Spin Columns, Thermo Scientific
Supplier: Thermo Scientific
D-(+)-Trehalose dihydrate 97.0-102.0% (by HPLC) NF, HPLE, multi-compendial parenteral grade, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Endothelial Cell Growth Media Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA