Order Entry
Canada
Orders LinkContactUsLinkComponent
RNA Binding Buffer, Zymo Research

RNA Binding Buffer, Zymo Research

50 mL
Catalog # 76020-602
Supplier:   Zymo Research
Supplier #:  R1013-2-50
RNA Binding Buffer, Zymo Research
Catalog # 76020-602
Supplier:  Zymo Research
CAS Number:  
RNA Binding Buffer, Zymo Research
RNA Binding Buffer, Zymo Research

50 mL

CAS Number:  
Price
$179.09
Log in for availability
Each (50ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Size:
    50 mL
  • Cat. No.:
    76020-602
  • Brand name:
    Zymo Research

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.