Order Entry
Canada
Orders LinkContactUsLinkComponent
Solution 3 Neutralizing Buffer, Zymo Research

Solution 3 Neutralizing Buffer, Zymo Research

20 mL
Catalog # 76020-312
Supplier:   Zymo Research
Supplier #:  D2004-3-20
Solution 3 Neutralizing Buffer, Zymo Research
Catalog # 76020-312
Supplier:  Zymo Research
CAS Number:  
Solution 3 Neutralizing Buffer, Zymo Research
Solution 3 Neutralizing Buffer, Zymo Research

20 mL

CAS Number:  
Price
$43.18
Log in for availability
Each (20ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Size:
    20 mL
  • Cat. No.:
    76020-312
  • Brand name:
    Zymo Research

Specifications