Order Entry
Canada
Orders LinkContactUsLinkComponent
Solution 3 Neutralizing Buffer, Zymo Research

Solution 3 Neutralizing Buffer, Zymo Research

180 mL
Catalog # 76020-310
Supplier:   Zymo Research
Supplier #:  D2004-3-180
Solution 3 Neutralizing Buffer, Zymo Research
Catalog # 76020-310
Supplier:  Zymo Research
CAS Number:  
Solution 3 Neutralizing Buffer, Zymo Research
Solution 3 Neutralizing Buffer, Zymo Research

180 mL

CAS Number:  
Price
$337.83
Log in for availability
Each (180ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Size:
    180 mL
  • Cat. No.:
    76020-310
  • Brand name:
    Zymo Research

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.