Order Entry
Canada
ContactUsLinkComponent
Reservoir 25ml
Polystyrene Reagent Reservoirs
Catalog # 470228-450
Supplier:  MTC BIO, INC.
undefined
Polystyrene Reagent Reservoirs
Catalog # 470228-450
Supplier:  MTC BIO, INC.
Supplier Number:  P8025_WARDS

Specifications

  • Sterility:
    Non sterile
  • Width:
    63 mm
  • Capacity:
    25 ml
  • Description:
    Reagent Reservoir
  • Height:
    27 mm
  • Length:
    150 mm
  • Cat. no.:
    470228-450

Specifications

About this item

Standard 4-channel, 8-channel, 12-channel and various special purpose pipettes are all compatible with these disposable, polystyrene reservoirs. To facilitate easy, dripless pouring, all versions have independent spouts in each corner. Bottoms are formed with a sharply tapered 'V' shape to minimize waste and reagent consumption.

  • Graduated
  • Anti-static
  • Sharp "V" Bottoms
  • Pouring spout on all 4 corners
  • Divided options are available
  • All reservoirs are RNAse- and DNAse-free

Optional Accessory

Product Information