Order Entry
Canada
Orders LinkContactUsLinkComponent
WARMSTART® RTx Reverse Transcriptase, New England Biolabs

WARMSTART® RTx Reverse Transcriptase, New England Biolabs

WarmStart RTx Reverse Transcriptase, 250, 250 Reactions
Catalog # CA102877-820
Supplier #:  M0380L
WARMSTART® RTx Reverse Transcriptase, New England Biolabs
Catalog # CA102877-820
CAS Number:  
WARMSTART® RTx Reverse Transcriptase, New England Biolabs
WARMSTART® RTx Reverse Transcriptase, New England Biolabs

WarmStart RTx Reverse Transcriptase, 250, 250 Reactions

CAS Number:  
Price
$486.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

WarmStart RTx Reverse Transcriptase is a unique in silico designed RNA-directed DNA polymerase coupled with a reversibly-bound aptamer that inhibits RTx activity below 40°C

This enzyme can synthesize a complementary DNA strand initiating from a primer using RNA (cDNA synthesis) or single-stranded DNA as a template. RTx is a robust enzyme for RNA detection in amplification reactions and is particularly well suited for use in LAMP (loop-mediated isothermal amplification). The WarmStart property enables high throughput applications, room temperature setup, and increases the consistency and specificity of amplification reactions. RTx contains intact RNase H activity.

Specifications

Specifications

  • Description:
    WarmStart RTx Reverse Transcriptase
  • Size:
    250 Reactions
  • No. of reactions:
    250

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.