Order Entry
Canada
Orders LinkContactUsLinkComponent
XhoI

XhoI

XhoI, 100000 units/ml, 25000 units
Catalog # CA101418-166
Supplier #:  R0146M
XhoI
Catalog # CA101418-166
CAS Number:  
XhoI
XhoI

XhoI, 100000 units/ml, 25000 units

CAS Number:  
Price
$491.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

An E.coli strain that carries the XhoI gene from Xanthomonas holcicola (ATCC 13461).

Specifications

Specifications

  • Concentration:
    100000 units/ml
  • Description:
    XhoI
  • Size:
    25000 units
  • Cat. No.:
    CA101418-166
  • Enzyme name:
    XhoI

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.