Order Entry
Canada
Orders LinkContactUsLinkComponent
NcoI

NcoI

NcoI, 10000 units/ml, 1000 units
Catalog # CA101228-984
Supplier #:  R0193S
NcoI
Catalog # CA101228-984
CAS Number:  
NcoI
NcoI

NcoI, 10000 units/ml, 1000 units

CAS Number:  
Price
$113.99
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

An E.coli strain that carries the NcoI gene from Nocardia corallina (ATCC 19070).

Specifications

Specifications

  • Concentration:
    10000 units/ml
  • Description:
    NcoI
  • Size:
    1000 units
  • Cat. No.:
    CA101228-984
  • Enzyme name:
    NcoI

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.