Orders LinkContactUsLinkComponent
USER
USER
250 units
Catalog # CA101228-398
Supplier #:  M5505L
USER
Catalog # CA101228-398
CAS Number:  
USER
USER
250 units
CAS Number:  
Price
$529.00
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
USER (Uracil-Specific Excision Reagent) Enzyme generates a single nucleotide gap at the location of a uracil
USER Enzyme is a mixture of Uracil DNA glycosylase (UDG) and the DNA glycosylase-lyase Endonuclease VIII. UDG catalyses the excision of a uracil base, forming an abasic (apyrimidinic) site while leaving the phosphodiester backbone intact. The lyase activity of Endonuclease VIII breaks the phosphodiester backbone at the 3´ and 5´ sides of the abasic site so that base-free deoxyribose is released.
Specifications
Specifications
  • Size:
    250 units
  • Cat. No.:
    CA101228-398
  • Enzyme name:
    User
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)