Orders LinkContactUsLinkComponent
Fpg
Fpg
2500 units
Catalog # CA101227-628
Supplier #:  M0240L
Fpg
Catalog # CA101227-628
CAS Number:  
Fpg
Fpg
2500 units
CAS Number:  
Price
$536.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Fpg (formamidopyrimidine [fapy]-DNA glycosylase), also known as 8-oxoguanine DNA glycosylase, acts both as an N-glycosylase and an AP-lyase
The N-glycosylase activity releases damaged purines from double-stranded DNA, generating an apurinic (AP) site. The AP-lyase activity cleaves both 3´ and 5´ to the AP site thereby removing the AP site and leaving a 1-base gap with a 5´ and 3´ phosphate. Some of the damaged bases recognized and removed by Fpg include 7, 8-dihydro-8-oxoguanine (8-oxoguanine), 8-oxoadenine, fapy-guanine, methy-fapy-guanine, fapy-adenine, aflatoxin B1-fapy-guanine, 5-hydroxy-cytosine and 5-hydroxy-uracil.
Specifications
Specifications
  • Size:
    2500 units
  • Cat. No.:
    CA101227-628
  • Enzyme name:
    Fpg